Merck
  • Correction of metabolic acidosis with potassium citrate in renal transplant patients and its effect on bone quality.
Correction of metabolic acidosis with potassium citrate in renal transplant patients and its effect on bone quality.
Clinical journal of the American Society of Nephrology : CJASN (2012-07-10)
Astrid Starke, Alf Corsenca, Thomas Kohler, Johannes Knubben, Marius Kraenzlin, Daniel Uebelhart, Rudolf P Wüthrich, Brigitte von Rechenberg, Ralph Müller, Patrice M Ambühl
ZUSAMMENFASSUNG
Acidosis and transplantation are associated with increased risk of bone disturbances. This study aimed to assess bone morphology and metabolism in acidotic patients with a renal graft, and to ameliorate bone characteristics by restoration of acid/base homeostasis with potassium citrate. This was a 12-month controlled, randomized, interventional trial that included 30 renal transplant patients with metabolic acidosis (S-[HCO(3)(-)] <24 mmol/L) undergoing treatment with either potassium citrate to maintain S-[HCO(3)(-)] >24 mmol/L, or potassium chloride (control group). Iliac crest bone biopsies and dual-energy X-ray absorptiometry were performed at baseline and after 12 months of treatment. Bone biopsies were analyzed by in vitro micro-computed tomography and histomorphometry, including tetracycline double labeling. Serum biomarkers of bone turnover were measured at baseline and study end. Twenty-three healthy participants with normal kidney function comprised the reference group. Administration of potassium citrate resulted in persisting normalization of S-[HCO(3)(-)] versus potassium chloride. At 12 months, bone surface, connectivity density, cortical thickness, and cortical porosity were better preserved with potassium citrate than with potassium chloride, respectively. Serological biomarkers and bone tetracycline labeling indicate higher bone turnover with potassium citrate versus potassium chloride. In contrast, no relevant changes in bone mineral density were detected by dual-energy X-ray absorptiometry. Treatment with potassium citrate in renal transplant patients is efficient and well tolerated for correction of metabolic acidosis and may be associated with improvement in bone quality. This study is limited by the heterogeneity of the investigated population with regard to age, sex, and transplant vintage.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumcitrat tribasisch Monohydrat, ≥98% (GC/titration)
Sigma-Aldrich
Kaliumcitrat Monohydrat, purum p.a., ≥99.0% (NT)
Sigma-Aldrich
Kaliumcitrat Monohydrat, meets analytical specification of Ph. Eur., BP, FCC, E332, 99-100.5% (calc with ref. to anhyd. subst.)
Sigma-Aldrich
Kaliumcitrat Monohydrat, meets USP testing specifications
Sigma-Aldrich
Kaliumcitrat tribasisch Monohydrat, suitable for cell culture