Merck
  • Influence of shade and light-curing distance on the degree of conversion and flexural strength of a dual-cure core build-up resin composite.
Influence of shade and light-curing distance on the degree of conversion and flexural strength of a dual-cure core build-up resin composite.
American journal of dentistry (2012-07-12)
Farid S El-Askary, Dalia I El-Korashy
ZUSAMMENFASSUNG
To investigate the influence of shade and light-curing distance on the degree of conversion (DC) and flexural strength (FS) of a dual-cure core build-up resin composite. 135 specimens were prepared and divided into three main groups according to the shade of the composite core material; dentin, blue and white shades. Each group was further subdivided into three subgroups according to the light-curing distance; 0 mm, 3 mm and 5 mm. The DC was measured using FTIR spectrometer. For testing the FS, the mini flexural test specimens were prepared and subjected to three point bending test in a universal testing machine. The results were analyzed using two-way and one-way ANOVA followed by Duncan's Multiple Range Test. The significance level was set at P = 0.05. Both the DC and FS of the dual-cure core build-up material used in this study were influenced by the shade of the material and the light-curing distance. The blue shade exhibited the highest percentage DC values while the white shade revealed the lowest percentage DC. The dentin shade at 0 mm light-curing distance exhibited the highest mean FS value; however the 3 mm and 5 mm light-curing distance of the white shade showed the lowest mean FS values.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triethylenglycoldimethacrylat, contains 80-120 ppm MEHQ as inhibitor, 95%
Sigma-Aldrich
Triethylenglycoldimethacrylat, contains 200 ppm monomethyl ether hydroquinone as inhibitor, 99%