Merck
  • Influence of anionic cosolutes and pH on nanoscale zerovalent iron longevity: time scales and mechanisms of reactivity loss toward 1,1,1,2-tetrachloroethane and Cr(VI).
Influence of anionic cosolutes and pH on nanoscale zerovalent iron longevity: time scales and mechanisms of reactivity loss toward 1,1,1,2-tetrachloroethane and Cr(VI).
Environmental science & technology (2012-07-12)
Yang Xie, David M Cwiertny
ZUSAMMENFASSUNG
Nanoscale zerovalent iron (NZVI) was aged over 30 days in suspension (2 g/L) with different anions (chloride, perchlorate, sulfate, carbonate, nitrate), anion concentrations (5, 25, 100 mN), and pH (7, 8). During aging, suspension samples were reacted periodically with 1,1,1,2-tetrachloroethane (1,1,1,2-TeCA) and Cr(VI) to determine the time scales and primary mode of NZVI reactivity loss. Rate constants for 1,1,1,2-TeCA reduction in Cl(-), SO(4)(2-), and ClO(4)(-) suspensions decreased by 95% over 1 month but were generally equivalent to one another, invariant of concentration and independent of pH. In contrast, longevity toward 1,1,1,2-TeCA depended upon NO(3)(-) and HCO(3)(-) concentration, with complete reactivity loss over 1 and 14 days, respectively, in 25 mN suspensions. X-ray diffraction suggests that reactivity loss toward 1,1,1,2-TeCA in most systems results from Fe(0) conversion into magnetite, whereas iron carbonate hydroxide formation limits reactivity in HCO(3)(-) suspensions. Markedly different trends in Cr(VI) removal capacity (mg Cr/g NZVI) were observed during aging, typically exhibiting greater longevity and a pronounced pH-dependence. Notably, a strong linear correlation exists between Cr(VI) removal capacities and rates of Fe(II) production measured in the absence of Cr(VI). While Fe(0) availability dictates longevity toward 1,1,1,2-TeCA, this correlation suggests surface-associated Fe(II) species are primarily responsible for Cr(VI) reduction.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,1,1,2-Tetrachlorethan, ReagentPlus®, 99%
Sigma-Aldrich
Ethan, ≥99%