Merck
  • Effect of diflubenzuron on the development of Pinus pinaster seedlings inoculated with the ectomycorrhizal fungus Pisolithus tinctorius.
Effect of diflubenzuron on the development of Pinus pinaster seedlings inoculated with the ectomycorrhizal fungus Pisolithus tinctorius.
Environmental science and pollution research international (2012-07-12)
Miguel A Ramos, Nadine R Sousa, Albina R Franco, Vítor Costa, Rui S Oliveira, Paula M L Castro
ZUSAMMENFASSUNG
Diflubenzuron (DFB) is an insecticide commonly used to control forest pests. The objectives of this study were to assess the effect of diflubenzuron on the development of Pinus pinaster seedlings and Pisolithus tinctorius under laboratory conditions and to study the possible protective role of this ectomycorrhizal fungus against the effects of diflubenzuron. In vitro experiments revealed that diflubenzuron inhibited fungal growth at all tested concentrations (0.01, 0.1, 1, 10 and 100 mg L(-1)). Root growth was inhibited at the two highest diflubenzuron concentrations. The activity of the antioxidant defence system of non-inoculated P. pinaster increased at 1 and 10 mg DFB kg(-1) substrate, and inoculation increased the threshold to the highest concentration. The protective role of the ectomycorrhizal fungus was seen in the increase of CAT activity. This study revealed that despite causing no mortality, diflubenzuron has the ability to cause sub-lethal damage to P. pinaster. The disproportionate use of this insecticide may lead to higher amounts of its residues in soil and the biosphere, endangering trees, fungi and their symbiosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Diflubenzuron, PESTANAL®, analytical standard