Merck
  • Formation of diclofenac molecular ions as the effect of Cu(2+)-π interaction under electrospray ionization mass spectrometry conditions.
Formation of diclofenac molecular ions as the effect of Cu(2+)-π interaction under electrospray ionization mass spectrometry conditions.
European journal of mass spectrometry (Chichester, England) (2012-07-17)
Wojciech Ostrowski, Katarzyna Linko, Rafał Frański
ZUSAMMENFASSUNG
Solutions containing diclofenac (M) and a copper salt [CuCl2, Cu(ClO4)2, Cu(NO3)2, CuSO4) were analysed by electrospray ionisation mass spectrometry (ESI-MS). Because of the cation-pi interactions in diclofenac-Cu(II) complexes, the diclofenac molecular ion M+* at m/z 295 was formed. It was found that the solvent composition (methanol versus water/methanol) and counter ion strongly affect the M+* ion formation. Formation pathways of ion M+* are discussed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kupfer(II)-chlorid Dihydrat, ACS reagent, ≥99.0%
Sigma-Aldrich
Kupfer(II)-chlorid, 97%
Sigma-Aldrich
Kupfer(II)-chlorid Dihydrat, reagent grade
Sigma-Aldrich
Kupfer(II)-chlorid, powder, 99%
Sigma-Aldrich
Kupfer(II)-chlorid, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Copper(II) chloride dihydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kupfer(II)-chlorid, 99.999% trace metals basis
Sigma-Aldrich
Kupfer(II)-chlorid Dihydrat, 99.999%
Sigma-Aldrich
Kupfer(II)-chlorid Dihydrat, ≥99.95% trace metals basis