Merck
Tandem transformation of glycerol to esters.
Journal of biotechnology (2012-07-17)
Maria V Sotenko, Martin Rebroš, Victor S Sans, Konstantin N Loponov, Matthew G Davidson, Gill Stephens, Alexei A Lapkin
ZUSAMMENFASSUNG
Tandem transformation of glycerol via microbial fermentation and enzymatic esterification is presented. The reaction can be performed with purified waste glycerol from biodiesel production in a continuous mode, combining continuous fermentation with membrane-supported enzymatic esterification. Continuous anaerobic fermentation was optimized resulting in the productivity of 2.4 g L⁻¹ h⁻¹ of 1,3-propanediol. Biphasic esterification of 1,3-propanediol was optimized to achieve ester yield of up to 75%. A hollow fibre membrane contactor with immobilized Rhizomucor miehei lipase was demonstrated for the continuous tandem fermentation-esterification process.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,3-Propandiol, 98%