Merck
  • T-2 toxin enhances catabolic activity of hypertrophic chondrocytes through ROS-NF-κB-HIF-2α pathway.
T-2 toxin enhances catabolic activity of hypertrophic chondrocytes through ROS-NF-κB-HIF-2α pathway.
Toxicology in vitro : an international journal published in association with BIBRA (2012-07-18)
Juan Tian, Jidong Yan, Wei Wang, Nannan Zhong, Lifang Tian, Jian Sun, Zixin Min, Jie Ma, Shemin Lu
ZUSAMMENFASSUNG
T-2 toxin (T-2), one of the most important and toxic trichothecene mycotoxins, can cause many medical problems, such as diarrhea, nervous disorders, immunodepression and death, and is also believed as an etiological factor of Kashin-Beck disease, an endemic osteochondropathy prevailing in North China. However, the molecular mechanisms underlying T-2 effects on tissue damage remain elusive. We differentiated ATDC5 chondrogenic cells into hypertrophic chondrocytes, and found that T-2 reduced the expression of anabolic genes, and increased the expression of catabolic genes. To uncover the mechanism that T-2 influenced metabolic homeostasis of hypertrophic chondrocytes, we observed that T-2 increased the production of reactive oxygen species (ROS) and the degradation of IκB-α, and up-regulated the expression of hypoxia-induced factor-2α (HIF-2α). Bay11-7085 (an inhibitor of NF-κB pathway) inhibited the up-regulation of HIF-2α, and N-acetyl-l-cysteine (a ROS scavenger) inhibited both the decrease of IκB-α and the up-regulation of HIF-2α. Our results demonstrate that ROS-NF-κB-HIF-2α pathway participates in the effects of T-2 on hypertrophic chondrocytes, and HIF-2α plays an important role as a key mediator in this process.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
T-2-Toxin, from Fusarium sp., powder, ≥98% (HPLC)
Supelco
T2-Toxin -Lösung, 100 μg/mL in acetonitrile, analytical standard