Merck
  • Biochemical properties of a novel and highly thermostable bacterial α-carbonic anhydrase from Sulfurihydrogenibium yellowstonense YO3AOP1.
Biochemical properties of a novel and highly thermostable bacterial α-carbonic anhydrase from Sulfurihydrogenibium yellowstonense YO3AOP1.
Journal of enzyme inhibition and medicinal chemistry (2012-07-19)
Clemente Capasso, Viviana De Luca, Vincenzo Carginale, Raffaele Cannio, Mosè Rossi
ZUSAMMENFASSUNG
A new carbonic anhydrase (CA, EC 4.2.1.1) from the thermophilic bacterium Sulfurihydrogenibium yellowstonense YO3AOP1 was identified and characterized. The bacterial carbonic anhydrase gene was expressed in Escherichia coli yielding an active enzyme, which was purified in large amounts. The recombinant protein (SspCA) was found to belong to the α-CA class and displays esterase activity. The kinetic parameters were determined by using CO(2) and p-nitrophenylacetate (p-NpA) as substrates. The bacterial enzyme presented specific activity comparable to that of bovine carbonic anhydrase (bCA II) but it showed biochemical properties never observed for the mammalian enzyme. The thermophilic enzyme, in fact, was endowed with high thermostability and with unaltered residual activity after prolonged exposure to heat up to 100°C. SspCA and the bovine carbonic anhydrase (bCA II) were immobilized within a polyurethane (PU) foam. The immobilized bacterial enzyme was found to be active and stable at 100°C up to 50 h.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Nitrophenylacetat, esterase substrate