Merck
  • Rosmarinus officinalis L. essential oil and the related compound 1,8-cineole do not induce direct or cross-protection in Listeria monocytogenes ATCC 7644 cultivated in meat broth.
Rosmarinus officinalis L. essential oil and the related compound 1,8-cineole do not induce direct or cross-protection in Listeria monocytogenes ATCC 7644 cultivated in meat broth.
Canadian journal of microbiology (2012-07-20)
Nelson Justino Gomes Neto, Isabelle Silva Luz, Wanessa Gonçalves Honório, Adassa Gama Tavares, Evandro Leite de Souza
ZUSAMMENFASSUNG
Listeria monocytogenes has the capability of adapting to 1 or more antimicrobial compounds or procedures applied by the food industry to control the growth and survival of microorganisms in foods. In this study, the effects of Rosmarinus officinalis essential oil (EO) and the related compound 1,8-cineole on the inhibition of the growth and survival of L. monocytogenes ATCC 7644 were determined. The ability of the R. officinalis EO and 1,8-cineole to induce direct and cross-protection of bacteria against various stresses (lactic acid, pH 5.2; NaCl, 3 g/100 mL; high temperature, 45 °C) was also determined. At all concentrations tested (minimum inhibitory concentration (MIC), ½ MIC, and ¼ MIC), both compounds inhibited the cell viability of L. monocytogenes over 120 min of exposure. Overnight exposure of L. monocytogenes to sublethal amounts of either the R. officinalis EO or 1,8-cineole in meat broth revealed no induction of direct or cross-protection against lactic acid, NaCl, or high temperature. Similarly, cells subjected to 24 h cycles of adaptation with increasing amounts (½ MIC to 2× MIC) of the EO and 1,8-cineole showed no increase in direct tolerance, as they were able to survive in growth medium containing up to ½ MIC of either substance. These results show the antimicrobial efficacy of R. officinalis EO and 1,8-cineole for use in systems, particularly as anti-L. monocytogenes compounds.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Eucalyptol, 99%
Sigma-Aldrich
Eucalyptol, natural, ≥99%, FCC, FG
Supelco
Eucalyptol, analytical standard
Supelco
1,8-Cineol, primary reference standard
Sigma-Aldrich
Eucalyptol, tested according to Ph. Eur.
Cineol, European Pharmacopoeia (EP) Reference Standard