Merck
  • Identification and quantification of 1-hydroxybutene-2-yl mercapturic acid in human urine by UPLC- HILIC-MS/MS as a novel biomarker for 1,3-butadiene exposure.
Identification and quantification of 1-hydroxybutene-2-yl mercapturic acid in human urine by UPLC- HILIC-MS/MS as a novel biomarker for 1,3-butadiene exposure.
Chemical research in toxicology (2012-07-24)
Katharina Sterz, Gerhard Scherer, Jan Krumsiek, Fabian J Theis, Josef Ecker
ZUSAMMENFASSUNG
1,3-Butadiene (BD) is a Class 1 carcinogen present at workplaces, in polluted air, in automobile exhaust, and in tobacco smoke. 2-Hydroxybutene-1-yl mercapturic acid (2-MHBMA) is a urinary metabolite often measured as a biomarker for exposure to BD. Here, we show for the first time that an additional MHBMA isomer is present at significant amounts in human urine, 1-hydroxybutene-2-yl mercapturic acid (1-MHBMA). For its quantification, a highly sensitive UPLC-HILIC-MS/MS method was developed and validated. Analyzing urinary samples of 183 volunteers, we demonstrate that 1-MHBMA is a novel and potentially more reliable biomarker for BD exposure than the commonly analyzed 2-MHBMA.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,3-Butadien -Lösung, 20 wt. % in toluene
Sigma-Aldrich
1,3-Butadien -Lösung, 15 wt. % in hexane