Merck
  • Study on the subgel-phase formation using an asymmetric phospholipid bilayer membrane by high-pressure fluorometry.
Study on the subgel-phase formation using an asymmetric phospholipid bilayer membrane by high-pressure fluorometry.
Langmuir : the ACS journal of surfaces and colloids (2012-07-25)
Masaki Goto, Agnieszka Wilk, Kosuke Kataoka, Shirish Chodankar, Nobutake Tamai, Makoto Fukui, Joachim Kohlbrecher, Hiro-O Ito, Hitoshi Matsuki
ZUSAMMENFASSUNG
The myristoylpalmitoylphosphatidylcholine (MPPC) bilayer membrane shows a complicated temperature-pressure phase diagram. The large portion of the lamellar gel (L(β)'), ripple gel (P(β)'), and pressure-induced gel (L(β)I) phases exist as metastable phases due to the extremely stable subgel (L(c)) phase. The stable L(c) phase enables us to examine the properties of the L(c) phase. The phases of the MPPC bilayers under atmospheric and high pressures were studied by small-angle neutron scattering (SANS) and fluorescence spectroscopy using a polarity-sensitive fluorescent probe Prodan. The SANS measurements clearly demonstrated the existence of the metastable L(β)I phase with the smallest lamellar repeat distance. From a second-derivative analysis of the fluorescence data, the line shape for the L(c) phase under high pressure was characterized by a broad peak with a minimum of ca. 460 nm. The line shapes and the minimum intensity wavelength (λ″(min)) values changed with pressure, indicating that the L(c) phase has highly pressure-sensible structure. The λ″(min) values of the L(c) phase spectra were split into ca. 430 and 500 nm in the L(β)I phase region, which corresponds to the formation of a interdigitated subgel L(c) (L(c)I) phase. Moreover, the phase transitions related to the L(c) phase were reversible transitions under high pressure. Taking into account the fluorescence behavior of Prodan for the L(c) phase, we concluded that the structure of the L(c) phase is highly probably a staggered structure, which can transform into the L(c)I phase easily.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Dimethyl-6-propionyl-2-naphthylamin, BioReagent, suitable for fluorescence, ≥98.0% (HPLC)