Merck
  • Characterization of catechol 2,3-dioxygenase from Planococcus sp. strain S5 induced by high phenol concentration.
Characterization of catechol 2,3-dioxygenase from Planococcus sp. strain S5 induced by high phenol concentration.
Acta biochimica Polonica (2012-07-25)
Katarzyna Hupert-Kocurek, Urszula Guzik, Danuta Wojcieszyńska
ZUSAMMENFASSUNG
This study aimed at characterization of a new catechol 2,3-dioxygenase isolated from a Gram-positive bacterium able to utilize phenol as the sole carbon and energy source. Planococcus sp. strain S5 grown on 1 or 2 mM phenol showed activity of both a catechol 1,2- and catechol 2,3-dioxygenase while at a higher concentrations of phenol only catechol 2,3-dioxygenase activity was observed. The enzyme was optimally active at 60°C and pH 8.0. Kinetic studies showed that the K(m) and V(max) of the enzyme were 42.70 µM and 329.96 mU, respectively. The catechol 2,3-dioxygenase showed the following relative meta-cleavage activities for various catechols tested: catechol (100%), 3-methylcatechol (13.67%), 4-methylcatechol (106.33%) and 4-chlorocatechol (203.80%). The high reactivity of this enzyme towards 4-chlorocatechol is different from that observed for other catechol 2,3-dioxygenases. Nucleotide sequencing and homology search revealed that the gene encoding the S5 catechol 2,3-dioxygenase shared the greatest homology with the known genes encoding isoenzymes from Gram-negative Pseudomonas strains.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Methylcatechin, ≥95%
Sigma-Aldrich
3-Methylcatechin, 98%
Sigma-Aldrich
4-Chlorkatechol, 97%