Merck
  • Halitosis in cystinosis patients after administration of immediate-release cysteamine bitartrate compared to delayed-release cysteamine bitartrate.
Halitosis in cystinosis patients after administration of immediate-release cysteamine bitartrate compared to delayed-release cysteamine bitartrate.
Molecular genetics and metabolism (2012-07-27)
Martine Besouw, Albert Tangerman, Elisabeth Cornelissen, Patrice Rioux, Elena Levtchenko
ZUSAMMENFASSUNG
Halitosis due to dimethylsulfide (DMS) generation is a major side effect of cysteamine in the treatment of cystinosis. Recently, an enteric coated formulation of cysteamine bitartrate (RP103) administered twice daily was demonstrated to be non-inferior for lowering WBC cystine levels compared to the non-enteric coated formulation (Cystagon®), administered 4 times per day. Since both formulations had different pharmacokinetic profiles, we compared DMS breath levels after administration of either RP103 or Cystagon® in four cystinosis patients. Although cysteamine areas under the curve (AUCs) were comparable, AUC of DMS was lower after the administration of RP103 compared to Cystagon®. This observation is of importance in cystinosis patients, since halitosis hampers compliance with cysteamine therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cysteamin -hydrochlorid, ≥98% (titration)
Sigma-Aldrich
Cysteamin, ~95%
Sigma-Aldrich
Dimethylsulfid, ≥99%, FCC, FG
Sigma-Aldrich
Cysteamin, ≥98.0% (RT)
Sigma-Aldrich
Dimethylsulfid, ≥99%
Sigma-Aldrich
Cysteamin -hydrochlorid, BioXtra
Sigma-Aldrich
Dimethylsulfid, anhydrous, ≥99.0%
Sigma-Aldrich
Dimethylsulfid, natural, ≥99%, FCC, FG
Sigma-Aldrich
Cysteamin -hydrochlorid, ≥97.0% (RT)
Supelco
Dimethylsulfid, analytical standard