Merck
Kinetic and computational evidence for an intermediate in the hydrolysis of sulfonate esters.
Organic & biomolecular chemistry (2012-07-27)
Ann C Babtie, Marcelo F Lima, Anthony J Kirby, Florian Hollfelder
ZUSAMMENFASSUNG
The hydrolytic reactions of sulfonate esters have previously been considered to occur by concerted mechanisms. We now report the observation of a break in a Brønsted correlation for the alkaline hydrolysis of aryl benzenesulfonates. On either side of a break-point, β(leaving group) values of -0.27 (pK(a) < 8.5) and -0.97 (pK(a) > 8.5) are measured. These data are consistent with a two-step mechanism involving a pentavalent intermediate that is also supported by QM/MM calculations. The emerging scenario can be explained by the combined effect of a strong nucleophile with a poor leaving group that compel a usually concerted reaction to favour a stepwise process.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumhydroxid, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Kaliumhydroxid, reagent grade, 90%, flakes
Sigma-Aldrich
Kalilauge, 45 wt. % in H2O
Sigma-Aldrich
Kaliumhydroxid, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Benzolsulfonsäure, 98.0% (T)
Sigma-Aldrich
Kaliumhydroxid, ≥85% KOH basis, pellets, white
Supelco
Kalilauge, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Kaliumhydroxid, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Kaliumhydroxid, technical, ≥85%, powder
Sigma-Aldrich
Natrium-benzolsulfonat, 97%
Supelco
Kaliumhydroxid-Konzentrat, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Benzolsulfonsäure, technical grade, 90%
Sigma-Aldrich
Kaliumhydroxid, tested according to Ph. Eur.
Sigma-Aldrich
Kaliumhydroxid, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets