Merck
The photodegradation of quercetin: relation to oxidation.
Molecules (Basel, Switzerland) (2012-07-28)
Stefano Dall'Acqua, Giorgia Miolo, Gabbriella Innocenti, Sergio Caffieri
ZUSAMMENFASSUNG
The photostability of quercetin in alcoholic solutions was studied. Both UVA and UVB light induced degradation of quercetin, yielding a single product 1 deriving from oxidation and addition of an alcohol molecule to the 2,3 double bond. The same mechanism operated when quercetin was dissolved in alkaline solutions, and again a product 2 due to oxidation and addition of water was characterized. Comparison with quercetin analogs confirmed that, despite the presence of five hydroxy groups in quercetin, those in positions 3, 3', and 4' are mainly involved in the antioxidant activity of the compound , as well as in its photolability.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Galangin, autophagy inducing flavonoid