- Synthesis from D-altrose of (5R,6R,7R,8S)-5,7-dihydroxy-8-hydroxymethylconidine and 2,4-dideoxy-2,4-imino-D-glucitol, azetidine analogues of swainsonine and 1,4-dideoxy-1,4-imino-D-mannitol.
Synthesis from D-altrose of (5R,6R,7R,8S)-5,7-dihydroxy-8-hydroxymethylconidine and 2,4-dideoxy-2,4-imino-D-glucitol, azetidine analogues of swainsonine and 1,4-dideoxy-1,4-imino-D-mannitol.
Organic letters (2012-07-31)
Noelia Araújo, Sarah F Jenkinson, R Fernando Martínez, Andreas F G Glawar, Mark R Wormald, Terry D Butters, Shinpei Nakagawa, Isao Adachi, Atsushi Kato, Akihide Yoshihara, Kazuya Akimitsu, Ken Izumori, George W J Fleet
PMID22839435
ZUSAMMENFASSUNG
Ring closure of a 3,5-di-O-triflate derived from D-altrose with benzylamine allowed the formation of both monocyclic and bicyclic azetidine analogues of swainsonine.