Merck
  • Effectiveness of cerium nitrate-silver sulfadiazine in the treatment of facial burns: a multicenter, randomized, controlled trial.
Effectiveness of cerium nitrate-silver sulfadiazine in the treatment of facial burns: a multicenter, randomized, controlled trial.
Plastic and reconstructive surgery (2012-07-31)
Irma M M H Oen, Margriet E van Baar, Esther Middelkoop, Marianne K Nieuwenhuis
ZUSAMMENFASSUNG
The face is a very frequent site of burn injuries. This multicenter, randomized, controlled trial thus investigates the effectiveness of cerium nitrate-silver sulfadiazine in the treatment of facial burns compared with silver sulfadiazine. Adult patients with acute facial burns admitted to Dutch burn centers were randomized to treatment with either cerium nitrate-silver sulfadiazine or silver sulfadiazine. Primary outcome was need for surgery and time to wound healing. Aesthetic and functional outcome was assessed at 3, 6, and 12 months after burn. From March of 2006 until January of 2009, 179 patients were randomized and 154 could be included. The two groups of patients (cerium nitrate-silver sulfadiazine group, n=78; silver sulfadiazine group, n=76), were comparable regarding sex, age, percentage total body surface area burned, and cause. During admission, four patients died, leaving 77 and 73 patients for primary analyses, respectively. Surgery was required in 13 (16.9 percent) compared with 15 patients (20.5 percent) (p=0.57; odds ratio, 0.8; 95 percent CI, 0.3 to 1.8), respectively. Median time to wound healing was 11.0 days in the cerium nitrate-silver sulfadiazine group (interquartile range, 7.0 to 15.0) and 9.0 days for silver sulfadiazine group (interquartile range, 5.0 to 15.75) (p=0.17). There were no significant differences in functional and aesthetic outcome. No differences were found in effectiveness of both treatments. The vast majority of facial burns do not require surgery, and treatment with cerium nitrate-silver sulfadiazine and silver sulfadiazine leads to satisfactory outcome, both aesthetically and functionally. Therapeutic, II.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Silber(I)-sulfadiazin, 98%