Merck
[Therapeutic drug monitoring of rufinamide].
Therapie (2012-08-02)
Danièle Bentué-Ferrer, Olivier Tribut, Marie-Clémence Verdier
ZUSAMMENFASSUNG
Rufinamide is a third-generation antiepileptic drug, available since early 2010 in France. It is indicated in combination therapy in the Lennox-Gastaut syndrome from the age of 4. It has orphan drug status. The bioavailability of rufinamide is high, but decreases with the dose and increases with food intake. Rufinamide is not metabolized by cytochromes but hydrolyzed by a carboxylesterase in an inactive carboxylic derivative. Elimination is mainly renal. The half-life varies from 6 to 10h. Although established from relatively few studies, exposure efficacy and exposure toxicity relationships are argued. A plasma concentration of 15 mg/L, obtained with a standard regimen, reduces the number of seizures of 25%. Few factors of intrinsic variability are described. There are few clinically significant pharmacokinetic interactions and they concern combinations with other antiepileptic drugs, especially valproate. Although there is no validated therapeutic range, the level of evidence for this therapeutic drug monitoring has been estimated at "possibly useful".
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Rufinamid, ≥98% (HPLC), powder