Merck
Chromatin staining of Drosophila testes.
Cold Spring Harbor protocols (2012-08-03)
Silvia Bonaccorsi, Maria G Giansanti, Giovanni Cenci, Maurizio Gatti
ZUSAMMENFASSUNG
This protocol describes chromatin staining of Drosophila testes. To visualize DNA, preparations fixed using methanol-acetone, paraformaldehyde, or formaldehyde can be stained with several DNA-binding dyes. If the slides are to be examined with a fluorescence microscope equipped with filters that permit ultraviolet (UV) excitation, suitable dyes for DNA staining are Hoechst 33258 or 4',6-diamidino-2-phenylindole (DAPI). If the slides are to be analyzed with a confocal microscope not equipped with a UV laser, DNA can be stained with either propidium iodide or TOTO-3 iodide.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hoechst 33258 -Lösung, 1 mg/mL in H2O, ≥98.0% (HPLC)
Sigma-Aldrich
bis-Benzimid H 33258, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC)
Sigma-Aldrich
bis-Benzimid H 33258, suitable for fluorescence, ≥98.0% (HPLC)
Sigma-Aldrich
bis-Benzimid H 33258, ≥98% (HPLC)