Merck
Production of biobutanediols by the hydrogenolysis of erythritol.
ChemSusChem (2012-08-07)
Yasushi Amada, Hideo Watanabe, Yuichirou Hirai, Yasuteru Kajikawa, Yoshinao Nakagawa, Keiichi Tomishige
ZUSAMMENFASSUNG
The hydrogenolysis of erythritol using an Ir-ReO(x)/SiO(2) catalyst was performed for the production of butanediols, which are widely used as a raw material of polymers. The activity and selectivity to butanediols on Ir-ReO(x)/SiO(2) was much higher than that on conventional hydrogenolysis catalysts. The maximum selectivity to 1,4- and 1,3-butanediols reached 33 and 12 % at 74 % conversion, respectively. The erythritol conversion and selectivity to butanediols was almost maintained during four repeating tests if small amounts of acid were added to the reaction and the catalyst was calcined again. The reaction kinetics, reactivity trends, and characterization results indicate a direct hydrogenolysis mechanism in which the hydride species on the Ir metal surface attacks the alkoxide species on the 3D ReO(x) clusters. Based on the production of erythritol by the fermentation of glucose and glycerol, erythritol hydrogenolysis may be a promising pathway for the production of biobutanediols.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
meso-Erythritol, ≥99% (GC)
Sigma-Aldrich
Ammoniumperrhenat, ≥99%
Sigma-Aldrich
Natriumperrhenat, 99.99% trace metals basis
Sigma-Aldrich
Kaliumperrhenat, 99%
Sigma-Aldrich
Ammoniumperrhenat, 99.999% trace metals basis
Sigma-Aldrich
Kaliumperrhenat, 99.98% trace metals basis