Merck
  • Preparation of magnetite/poly(styrene-divinylbenzene) nanoparticles for selective enrichment-determination of fenitrothion in environmental and biological samples.
Preparation of magnetite/poly(styrene-divinylbenzene) nanoparticles for selective enrichment-determination of fenitrothion in environmental and biological samples.
Analytica chimica acta (2012-08-14)
Habibollah Eskandari, Ali Naderi-Darehshori
ZUSAMMENFASSUNG
In the present study, a cross-linked nano-sized spherical magnetic poly(styrene-divinylbenzene) is synthesized and used as an adsorbent for enrichment-determination of fenitrothion. A miniemulsion polymerization procedure was used to prepare the adsorbent. The magnetic adsorbent was characterized by FT-IR, SEM and TEM. The prepared magnetic adsorbent nanoparticles were mixed with magnetite nanoparticles for faster and more efficient magnetic precipitation. The reduced fenitrothion was coupled with 3-methyl-2-benzothiazolinone hydrazone and then the blue colored complex was extracted. The blue derivative of fenitrothion was eluted by a 1 mL aliquot of 1-propanol prior to spectrophotometry at 571 nm. Beer's law was obeyed in the range of 2-230 ng mL(-1) of fenitrothion with relative standard deviation and recovery in the ranges of 0.9-5.1% and 97.2-100.0%, respectively. Selectivity of the method was evaluated, and the method was successfully applied to the determination of fenitrothion in various water, soil, urine and human plasma samples.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Divinylbenzol, technical grade, 55%
Sigma-Aldrich
Divinylbenzol, technical grade, 80%
Supelco
Fenitrothion, PESTANAL®, analytical standard