Merck
  • Successful pregnancy outcome after treatment with C1-inhibitor concentrate in a patient with hereditary angioedema and a history of four miscarriages.
Successful pregnancy outcome after treatment with C1-inhibitor concentrate in a patient with hereditary angioedema and a history of four miscarriages.
European journal of obstetrics, gynecology, and reproductive biology (2012-08-14)
Henriette Farkas, Dorottya Csuka, Ferenc Tóth, László Kőszegi, Lilian Varga
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Danazol, ≥98%