Merck
  • New spectrophotometric methods for the determinations of hydrogen sulfide present in the samples of lake water, industrial effluents, tender coconut, sugarcane juice and egg.
New spectrophotometric methods for the determinations of hydrogen sulfide present in the samples of lake water, industrial effluents, tender coconut, sugarcane juice and egg.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-08-15)
B Shyla, G Nagendrappa
ZUSAMMENFASSUNG
The new methods are working on the principle that iron(III) is reduced to iron(II) by hydrogen sulfide, catechol and p-toluidine the system 1/hydrogen sulfide the system 2, in acidic medium followed by the reduced iron forming complex with 1,10-phenanthroline with λ(max) 510 nm. The other two methods are based on redox reactions between electrolytically generated manganese(III) sulfate taken in excess and hydrogen sulfide followed by the unreacted oxidant oxidizing diphenylamine λ(max) 570 the system 3/barium diphenylamine sulphonate λ(max) 540 nm, the system 4. The increase/decrease in the color intensity of the dye products of the systems 1 and 2 or 3 and 4 are proportional to the concentration of hydrogen sulfide with its quantification range 0.035-1.40 μg ml(-1)/0.14-1.40 μg ml(-1).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Mangan(II)-Sulfat Monohydrat, ReagentPlus®, ≥99%
Sigma-Aldrich
Mangan(II)-Sulfat Monohydrat, ACS reagent, ≥98%
Sigma-Aldrich
p-Toluidin, 99.6%
Sigma-Aldrich
Mangan(II)-Sulfat Monohydrat, BioReagent, suitable for plant cell culture, suitable for cell culture
Sigma-Aldrich
p-Toluidin, 99%
Sigma-Aldrich
Mangan -sulfat Monohydrat, meets USP testing specifications
Sigma-Aldrich
p-Toluidin -hydrochlorid, purum, ≥99.0% (AT)
Supelco
p-Toluidin, for spectrophotometric det. of Au, Tl(III), W, ≥99.0%