Merck
  • Protein motions are coupled to the reaction chemistry in coenzyme B12-dependent ethanolamine ammonia lyase.
Protein motions are coupled to the reaction chemistry in coenzyme B12-dependent ethanolamine ammonia lyase.
Angewandte Chemie (International ed. in English) (2012-08-16)
Henry J Russell, Alex R Jones, Sam Hay, Gregory M Greetham, Michael Towrie, Nigel S Scrutton
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Coenzym B12, ≥97.0%