Merck
Optical studies on electron beam evaporated lithium triborate films.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-08-29)
R Mohandoss, S Dhanuskodi, C Sanjeeviraja
ZUSAMMENFASSUNG
Lithium triborate (LB3) has numerous applications in scintillator for neutron detection, laser weapon and communication. LB3 films have been prepared by electron beam evaporation technique under a pressure of 1×10(-5) mbar on glass substrate at 323 K for 4 min. The crystallographic orientations and the lattice parameters (a=8.55 (2); b=5.09 (2); c=7.39 (2)Å) were determined by powder XRD indicating the (111) preferential orientation of the film. The lower cut off wavelength at 325 nm with 75% transparency was measured from the UV-vis spectrum. The optical constants extinction coefficient (K), reflectance (R), the linear refractive index (1.34) and the optical energy band gap (~4.0 eV) were estimated. The photoluminescence spectrum shows the emission peak in the visible region with low concentration of oxygen defects. LB3 is found to be second harmonic generation (SHG) active using a Q-switched Nd:YAG laser (1064 nm, 9 ns, 10 Hz). The nonlinear refractive index (n(2)~10(-16) cm(2)/W) and nonlinear absorption coefficient (β~10(-2) cm/W) reveal (Z-scan technique) that the material has negative nonlinearity and self-focusing nature.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lithiumtetraborat, ≥99.9% trace metals basis
Sigma-Aldrich
Lithiummetaborat, 99.9% trace metals basis
Sigma-Aldrich
Lithiumtetraborat, ≥99.995% trace metals basis
Sigma-Aldrich
Lithiummetaborat, ACS reagent, ≥98.0%
Sigma-Aldrich
Lithiummetaborat, 99.995% trace metals basis