Merck
  • Fumigant toxicity of plant essential oils against Camptomyia corticalis (Diptera: Cecidomyiidae).
Fumigant toxicity of plant essential oils against Camptomyia corticalis (Diptera: Cecidomyiidae).
Journal of economic entomology (2012-08-30)
Jun-Ran Kim, Perumalsamy Haribalan, Bong-Ki Son, Young-Joon Ahn
ZUSAMMENFASSUNG
The toxicity of 98 plant essential oils against third instars of cecidomyiid gall midge Camptomyia corticalis (Loew) (Diptera: Cecidomyiidae) was examined using a vapor-phase mortality bioassay. Results were compared with that of a conventional insecticide dichlorvos. Based on 24-h LC50 values, all essential oils were less toxic than dichlorvos (LC50, 0.027 mg/cm3). The LC50 of caraway (Carum carvi L.) seed, armoise (Artemisia vulgaris L.), clary sage (Salvia sclarea L.), oregano (Origanum vulgare L.), lemongrass [Cymbopogon citratus (DC.) Stapf], niaouli (Melaleuca viridiflora Gaertner), spearmint (Mentha spicata L.), cassia especial (Cinnamomum cassia Nees ex Blume), Dalmatian sage (Salvia offcinalis L.), red thyme (Thymus vulgaris L.), bay [Pimenta racemosa (P. Mill.) J.W. Moore], garlic (Allium sativum L.), and pennyroyal (Mentha pulegium L.) oils is between 0.55 and 0.60 mg/cm3. The LC50 of cassia (C. cassia, pure and redistilled), white thyme (T. vulgaris), star anise (Illicium verum Hook.f.), peppermint (Mentha X piperita L.), wintergreen (Gaultheria procumbens L.), cinnamon (Cinnamomum zeylanicum Blume) bark, sweet marjoram (Origanum majorana L.), Roman chamomile [Chamaemelum nobile (L.) All.], eucalyptus (Eucalyptus globulus Labill.), rosemary (Rosmarinus officinalis L.),Virginian cedarwood (Juniperus virginiana L.), pimento berry [Pimenta dioica (L.) Merr.], summer savory (Satureja hortensis L.), lavender (Lavandula angustifolia Mill.), and coriander (Coriandrum sativum L.) oils is between 0.61 and 0.99 mg/cm3. All other essential oils tested exhibited low toxicity to the cecidomyiid larvae (LC50, >0.99 mg/cm3). Global efforts to reduce the level of highly toxic synthetic insecticides in the agricultural environment justify further studies on the active essential oils as potential larvicides for the control of C. corticalis populations as fumigants with contact action.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Dichlorvos, PESTANAL®, analytical standard