Merck
Effect of sertraline on [Ca2+](i) and viability of human MG63 osteosarcoma cells.
Drug and chemical toxicology (2012-08-31)
Ko-Long Lin, Chao-Chuan Chi, Ti Lu, Li-Ling Tseng, Jue-Long Wang, Yi-Chau Lu, Chung-Ren Jan
ZUSAMMENFASSUNG
The antidepressant, sertraline, has been shown to have diverse in vitro effects. This study examined whether sertraline altered [Ca(2+)](i) in MG63 human osteosarcoma cells by using fura-2 as a Ca(2+)-sensitive fluorescent dye. At 50-200 µM, sertraline induced a [Ca(2+)](i) rise in a concentration-dependent manner. Ca(2+) response was decreased by removing extracellular Ca(2+), suggesting that Ca(2+) entry and release contributed to the [Ca(2+)](i) signal. Sertraline-induced Ca(2+) entry was inhibited by nifedipine, La(3+), Gd(3+), and SK&F96365. When extracellular Ca(2+) was removed, pretreatment with the endoplasmic reticulum (ER) Ca(2+) pump inhibitor, thapsigargin, or 2,5-di-tert-butylhydroquinone (BHQ) abolished the sertraline-evoked [Ca(2+)](i) rise. Incubation with sertraline also abolished the thapsigargin or BHQ-induced [Ca(2+)](i) rise. Inhibition of phospholipase C (PLC) with U73122 abolished the sertraline-induced [Ca(2+)](i) rise. At 20-30 µM, overnight treatment with sertraline killed cells in a concentration-dependent manner. The cytotoxic effect of sertraline was not reversed by chelating cytosolic Ca(2+) with 1,2-bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid (BAPTA). Annexin V/propidium iodide staining data demonstrate that sertraline (30 µM) evoked apoptosis. Sertraline (20 and 30 µM) also increased levels of reactive oxygen species. Together, in human osteosarcoma cells, sertraline evoked a [Ca(2+)](i) rise by inducing PLC-dependent Ca(2+) release from the ER and Ca(2+) entry by L-type Ca(2+) channels and store-operated Ca(2+) channels. Sertraline induced cell death that may involve apoptosis by mitochondrial pathways.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,2-Bis(2-Aminophenoxy)ethan-N,N,NN,N,N′,N′-Tetraessigsäure, 98%
Sigma-Aldrich
2,5-Di-tert-butylhydrochinon, 99%
Sigma-Aldrich
1,2-Bis-(2-aminophenoxy)-ethan-N,N,N′,N′-tetraessigsäure, ≥96.0% (HPLC)