Merck
  • Interplay between desolvation and secondary structure in mediating cosolvent and temperature induced alpha-synuclein aggregation.
Interplay between desolvation and secondary structure in mediating cosolvent and temperature induced alpha-synuclein aggregation.
Physical biology (2012-08-31)
V L Anderson, W W Webb, D Eliezer
ZUSAMMENFASSUNG
Both increased temperature and moderate concentrations of fluorinated alcohols enhance aggregation of the Parkinson's disease-associated protein α-synuclein (αS). Here, we investigate the secondary structural rearrangements induced by heating and trifluoroethanol [TFE]. At low TFE concentrations, CD spectra feature a negative peak characteristic of disordered polypeptides near 200 nm and a slight shoulder around 220 nm suggesting some polyproline-II content. Upon heating, these peaks weaken, while a weak negative signal develops at 222 nm. At high TFE concentrations, the spectra show distinct minima at 208 and 222 nm, indicative of considerable α-helical structure, which diminish upon heating. We observe a crossover between the low-TFE and high-TFE behavior near 15% TFE, where we previously showed that a partially helical intermediate is populated. We postulate that the protein is well solvated by water at low TFE concentrations and by TFE at high TFE concentrations, but may become desolvated at the crossover point. We discuss the potential roles and interplay of desolvation and helical secondary structure in driving αS aggregation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,2,2-Trifluorethanol, ReagentPlus®, ≥99%
Supelco
2,2,2-Trifluorethanol, analytical standard, suitable for NMR (spectroscopy)