- Pd(II)-mediated cyclization of o-allylbenzaldehydes in water: a novel synthesis of isocoumarins.
Pd(II)-mediated cyclization of o-allylbenzaldehydes in water: a novel synthesis of isocoumarins.
Organic letters (2012-09-01)
Po-Yuan Chen, Keng-Shiang Huang, Chin-Chuan Tsai, Tzu-Pin Wang, Eng-Chi Wang
PMID22934626
ZUSAMMENFASSUNG
A novel, concise and efficient synthesis of substituted isocoumarins is disclosed. o-Allylbenzaldehydes prepared from isovanillin were mediated by PdCl(2)-CuCl(2) in water to undergo a domino reaction sequence, including 6-exo-trig cyclization, the addition of water, the elimination of PdHCl, the isomerization of carbon-carbon double bond, the oxidation of hemiacetals with the elimination of PdHCl, and regeneration of PdCl(2)in situ to yield a series of new substituted isocoumarins in high yields, in one pot.