- Aryl-linked imidazolidin-2-ones as non-peptidic β-strand mimetics.
Aryl-linked imidazolidin-2-ones as non-peptidic β-strand mimetics.
Chemical communications (Cambridge, England) (2012-09-01)
Charlotte L Sutherell, Sam Thompson, Richard T W Scott, Andrew D Hamilton
PMID22936296
ZUSAMMENFASSUNG
We describe the design and synthesis of a non-peptidic β-strand mimetic composed of alternating aryl and imidazolidin-2-one rings that can be adapted to display diverse side-chains. Solid- and solution-phase data together with calculations suggest that the desired conformation for side-chain mimicry is readily accessible and well-populated.
MATERIALIEN