- Five new 3,4-seco-lanostane-type triterpenoids with antiproliferative activity in human leukemia cells isolated from the roots of Kadsura coccinea.
Five new 3,4-seco-lanostane-type triterpenoids with antiproliferative activity in human leukemia cells isolated from the roots of Kadsura coccinea.
Planta medica (2012-09-06)
Nan Wang, Zhan-lin Li, Dan-dan Song, Wei Li, Yue-hu Pei, Yong-kui Jing, Hui-ming Hua
PMID22948612
ZUSAMMENFASSUNG
Five new 3,4-seco-lanostane-type triterpenoids, seco-coccinic acids G-K (1-5), and a known compound, seco-coccinic F, were isolated from the roots of Kadsura coccinea (Lem.) A. C. Sm. Their structures were elucidated by spectroscopic methods, including 2D-NMR and HR-MS techniques. The cell growth inhibitory effects of these compounds were assayed in human leukemia HL-60 cells, and it was found that 1, 5, and 6 showed antiproliferative effects with GI₅₀ values of 28.4, 15.2, and 16.6 µM, respectively.
MATERIALIEN