Merck
  • Volatiles from apple trees infested with light brown apple moth larvae attract the parasitoid Dolichogenidia tasmanica.
Volatiles from apple trees infested with light brown apple moth larvae attract the parasitoid Dolichogenidia tasmanica.
Journal of agricultural and food chemistry (2012-09-07)
D M Suckling, A M Twidle, A R Gibb, L M Manning, V J Mitchell, T E S Sullivan, S L Wee, A M El-Sayed
ZUSAMMENFASSUNG
The volatile compounds emitted from uninfested apple seedlings, cv. Royal Gala, and apple seedlings infested with generalist herbivore Epiphyas postvittana larvae were sampled using headspace collection and analyzed by gas chromatography-mass spectrometry. Nine additional compounds were only detected in infested apple seedlings [including benzyl alcohol, (E)-β-ocimene, benzyl cyanide, indole, (E)-nerolidol, and four unidentified compounds]. Infested apple seedlings produced larger amounts of (Z)-3-hexenyl acetate, linalool, 4,8-dimethyl-1,3(E),7-nonatriene, methyl salicylate, β-caryophyllene, germacrene D, (E,E)-α-farnesene, and (Z)-3-hexenyl benzoate than uninfested plants. Female parasitoids flew exclusively upwind to infested and not to uninfested apple seedlings in wind tunnel choice tests and preferred infested leaflets in still air, even after the removal of larvae. The attraction of a parasitoid to infested apple seedlings in the laboratory and in the field to apple and many other plants in at least six families supports considerable generality of the tritrophic signaling process.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Linalool, 97%
Sigma-Aldrich
Linalool, ≥97%, FCC, FG
Sigma-Aldrich
Ocimen, mixture of isomers, stabilized, ≥90%
Sigma-Aldrich
L-Linalool, natural, ≥95%, FG
Supelco
Linalool, analytical standard
Sigma-Aldrich
cis-3-Hexenylacetat, ≥98%, stabilized, FCC, FG
Sigma-Aldrich
Nerolidol, 98%, Mixture of cis and trans
Sigma-Aldrich
(−)-Linalool, ≥95.0% (sum of enantiomers, GC)
Sigma-Aldrich
Nerolidol, mixture of cis and trans, ≥97%, stabilized, FG
Supelco
trans-Nerolidol, analytical standard
Supelco
(−)-Linalool, analytical standard