Merck
  • Chemotherapy and radiotherapy combination extends survival for patients with oligodendroglial tumors.
Chemotherapy and radiotherapy combination extends survival for patients with oligodendroglial tumors.
Cancer (2012-09-07)
Carrie Printz
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lomustin, ≥98%
Sigma-Aldrich
Procarbazin -hydrochlorid, ≥98% (HPLC)