Merck
Apoptotic effects of cooked and in vitro digested soy on human prostate cancer cells.
Food chemistry (2012-09-08)
Xin Dong, Wenqing Xu, Robert A Sikes, Changqing Wu
ZUSAMMENFASSUNG
Previous laboratory and animal studies reported that soy isoflavones were major bioactive compounds in soy to exert chemoprotection of prostate cancer. However, these studies cannot reflect the realistic effects that soy may induce through diets, and little is known about the bioavailability of isoflavones from whole soy food and their bioactivities after cooking and digestion. In this study, cooking and in vitro digestion were used to prepare soy extracts and the effects of cooking and digestion on the isoflavone contents and bioactivities of the whole soy extracts were examined. The cooking procedure generally increased the amount of daidzin, genistin and daidzein, but decreased that of genistein. Digestion process significantly lowered contents of daidzin and genistin in 60min cooked sample, while increased the contents of daidzin and daidzein and decreased the content of genistein in the uncooked sample. Antioxidant activities of soy extracts increased after cooking and in vitro digestion, while no consistent increase of the four soy isoflavones was determined. The apoptotic effects of soy extracts on both LNCaP and C4-2B cells were generally in a dose-dependent manner. Compared to purified single isoflavones, cooked and digested soy were more effective on induction of prostate cancer cell apoptosis, which indicated synergistic interactions between various bioactive compounds in the whole soy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Daidzein, ≥98%, synthetic
Sigma-Aldrich
Daidzin, ≥95.0% (HPLC)
Sigma-Aldrich
Genistin, from Glycine max (soybean), ≥95% (HPLC)
Supelco
Daidzein, analytical standard
Supelco
Genistin, analytical standard
Supelco
Daidzin, analytical standard