Merck
  • Highly sensitive optical biosensor for thrombin based on structure switching aptamer-luminescent silica nanoparticles.
Highly sensitive optical biosensor for thrombin based on structure switching aptamer-luminescent silica nanoparticles.
Journal of fluorescence (2012-09-12)
Ethiraju Babu, Paulpandian Muthu Mareeswaran, Seenivasan Rajagopal
ZUSAMMENFASSUNG
We describe here the construction of a sensitive and selective optical sensor system for the detection of human α-thrombin. The surface functionalized luminescent [Ru(dpsphen)(3)](4-) (dpsphen-4,7-diphenyl-1,10-phenanthroline disulfonate) ion doped silica nanoparticles (SiNPs) with a size ~70 nm have been prepared. The DABCYL (2-(4-dimethylaminophenyl)diazenyl-benzoic acid) quencher labeled thrombin binding aptamer is conjugated to the surface of SiNPs using BS(3) (bis(sulfosuccinimidyl) suberate) as a cross-linker, resulting in the conformational change of aptamer to form G-quadruplex structure upon the addition of thrombin. The binding event is translated into a change in the luminescence intensity of Ru(II) complex via FRET mechanism, due to the close proximity of DABCYL quencher with SiNPs. The selective detection of thrombin using the SiNPs-aptamer system up to 4 nM is confirmed by comparing its sensitivity towards other proteins. This work demonstrates the application of simple aptamer-SiNPs conjugate as a highly sensitive system for the detection of thrombin and also it is highly sensitive towards thrombin in the presence of other proteins and complex medium such as BSA.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Buttergelb, analytical standard