Merck
Metabolic ability and individual characteristics of an atrazine-degrading consortium DNC5.
Journal of hazardous materials (2012-09-18)
Ying Zhang, Bo Cao, Zhao Jiang, Xiaonan Dong, Miao Hu, Zhigang Wang
ZUSAMMENFASSUNG
A stable four-member bacterial consortium, DNC5 that was capable of metabolizing atrazine was isolated from corn-planted soil. The main objective of this paper is to characterize the individual metabolic characteristics and the mutualism of the cultivable members in the consortium DNC5. Substrates utilizing character of each community member indicate that the primary organism in this consortium is Arthrobacter sp. DNS10, which was the only strain capable of mineralizing atrazine to cyanuric acid. Two secondary strains (Bacillus subtilis DNS4 and Variovorax sp. DNS12) utilized cyanuric acid during the atrazine degradation process. Meanwhile, we found that a metabolite (isopropylamine) inhibited the atrazine degrading species Arthrobacter sp. DNS10. The last strain (Arthrobacter sp. DNS9) of this consortium played a role in reducing this inhibition by utilizing isopropylamine for its growth. Altogether this is a new combination of isolates in an atrazine degrading consortium. The growth and the degradation rate of consortium DNC5 were faster than that of the single strain DNS10. The high degradation ability of the consortium showed good potential for atrazine biodegradation. This study will contribute toward a better understanding about metabolic activities of atrazine degrading consortium, which are generally considered to be responsible for atrazine mineralization in the natural environment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumnitrat, ACS reagent, ≥99.0%
Sigma-Aldrich
Kaliumnitrat, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Kaliumnitrat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Isopropylamin, ≥99.5%
Sigma-Aldrich
Kaliumnitrat, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Ethylamin -Lösung, 66.0-72.0% in H2O
Sigma-Aldrich
Ethylamin, 97%
Sigma-Aldrich
Kaliumnitrat, 99.999% trace metals basis
Sigma-Aldrich
Ethylamin -Lösung, 2.0 M in THF
Sigma-Aldrich
Ethylamin -Lösung, 2.0 M in methanol
Sigma-Aldrich
Kaliumnitrat, BioXtra, ≥99.0%
Sigma-Aldrich
Kaliumnitrat, 99.99% trace metals basis
Supelco
Mettler-Toledo-Kalibriersubstanz ME 51143095, Kaliumnitrat, traceable to primary standards (LGC)
Sigma-Aldrich
Isopropylamin, ≥97.0% (GC)
Supelco
Isopropylamin, anhydrous, analytical standard