Merck
Effect of a calcium-silicate-based restorative cement on pulp repair.
Journal of dental research (2012-09-18)
X V Tran, C Gorin, C Willig, B Baroukh, B Pellat, F Decup, S Opsahl Vital, C Chaussain, T Boukpessi
ZUSAMMENFASSUNG
In cases of pulp injury, capping materials are used to enhance tertiary dentin formation; Ca(OH)(2) and MTA are the current gold standards. The aim of this study was to evaluate the capacity of a new calcium-silicate-based restorative cement to induce pulp healing in a rat pulp injury model. For that purpose, cavities with mechanical pulp exposure were prepared on maxillary first molars of 27 six-week-old male rats, and damaged pulps were capped with either the new calcium-silicate-based restorative cement (Biodentine), MTA, or Ca(OH)(2). Cavities were sealed with glass-ionomer cement, and the repair process was assessed at several time-points. At day 7, our results showed that both the evaluated cement and MTA induced cell proliferation and formation of mineralization foci, which were strongly positive for osteopontin. At longer time-points, we observed the formation of a homogeneous dentin bridge at the injury site, secreted by cells displaying an odontoblastic phenotype. In contrast, the reparative tissue induced by Ca(OH)(2) showed porous organization, suggesting a reparative process different from those induced by calcium silicate cements. Analysis of these data suggests that the evaluated cement can be used for direct pulp-capping.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Calciumsilicat, purum, 12-22% Ca (as CaO) basis, ≥87% SiO2 basis
Sigma-Aldrich
Calciumsilikat, −200 mesh, ≥90%