- Spontaneous stone passage: is it Ammi visnaga effect?
Spontaneous stone passage: is it Ammi visnaga effect?
Urological research (2012-09-20)
Isa Kilicaslan, Selcuk Coskun
PMID22990409
ZUSAMMENFASSUNG
Ammi visnaga was used in Ancient Egypt as an herbal remedy for renal colic. "Khellin", a chemical obtained from Ammi visnaga, was used as a smooth muscle relaxant and has been thought to have pleiotropic effects on urolithiasis. We report a case with multiple ureteral stone passages possibly as a result of medication with an herb preparation, Khellin.
MATERIALIEN