Merck
Hydration properties of small hydrophobic molecules by Brillouin light scattering.
The Journal of chemical physics (2012-09-25)
L Comez, L Lupi, M Paolantoni, F Picchiò, D Fioretto
ZUSAMMENFASSUNG
We study the relaxation of water molecules next to hydrophobic solutes with different functional groups by Brillouin light scattering. Evidence is given for (i) water activation energy in trimethylamine-N-oxide, proline and t-butyl alcohol diluted solutions which is comparable to that of neat water, almost independent from solute mole fraction and (ii) moderate slowdown of relaxation time of proximal water compared to the bulk, which is consistent with excluded volume models. Assuming that the main contribution to viscosity comes from bulk and hydration water, a rationale is given of the phenomenological Arrhenius' laws for the viscosity of diluted aqueous solutions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
tert-Butanol, ACS reagent, ≥99.0%
Sigma-Aldrich
tert-Butanol, suitable for HPLC, ≥99.5%
Sigma-Aldrich
tert-Butanol, ≥99% (GC)
Sigma-Aldrich
tert-Butanol, anhydrous, ≥99.5%
Sigma-Aldrich
Trimethylamin-N-oxid, 95%
Sigma-Aldrich
tert-Butanol, TEBOL® 99, ≥99.3%
Supelco
tert-Butanol, analytical standard