Merck
  • Structural analysis of the pyroglutamate-modified isoform of the Alzheimer's disease-related amyloid-β using NMR spectroscopy.
Structural analysis of the pyroglutamate-modified isoform of the Alzheimer's disease-related amyloid-β using NMR spectroscopy.
Journal of peptide science : an official publication of the European Peptide Society (2012-09-25)
Na Sun, Rudolf Hartmann, Justin Lecher, Matthias Stoldt, Susanne Aileen Funke, Lothar Gremer, Hans-Henning Ludwig, Hans-Ulrich Demuth, Martin Kleinschmidt, Dieter Willbold
ZUSAMMENFASSUNG
The aggregation of the Aβ plays a fundamental role in the pathology of AD. Recently, N-terminally modified Aβ species, pE-Aβ, have been described as major constituents of Aβ deposits in the brains of AD patients. pE-Aβ has an increased aggregation propensity and shows increased toxicity compared with Aβ1-40 and Aβ1-42. In the present work, high-resolution NMR spectroscopy was performed to study pE-Aβ3-40 in aqueous TFE-containing solution. Two-dimensional TOCSY and NOESY experiments were performed. On the basis of NOE and chemical shift data, pE-Aβ3-40 was shown to contain two helical regions formed by residues 14-22 and 30-36. This is similar as previously described for Aβ1-40. However, the secondary chemical shift data indicate decreased helical propensity in pE-Aβ3-40 when compared with Aβ1-40 under exactly the same conditions. This is in agreement with the observation that pE-Aβ3-40 shows a drastically increased tendency to form β-sheet-rich structures under more physiologic conditions. Structural studies of pE-Aβ are crucial for better understanding the structural basis of amyloid fibril formation in the brain during development of AD, especially because an increasing number of reports indicate a decisive role of pE-Aβ for the pathogenesis of AD.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Pyroglutaminsäure, ≥99.0% (T)
Sigma-Aldrich
L-Pyroglutaminsäure, BioXtra