- Mesoporous nanomagnet supercaptors for selective heme-proteins from human cells.
Mesoporous nanomagnet supercaptors for selective heme-proteins from human cells.
Chemical communications (Cambridge, England) (2012-09-28)
Mohamed Khairy, Sherif A El-Safty, Mohamed Ismael
PMID23014515
ZUSAMMENFASSUNG
Nanomagnet-selective supercaptors of heme-proteins (iron-porphyrin prosthetic group) based on mesoporous NiO and Fe(3)O(4) NPs were fabricated.