Merck
  • Facile fabrication of SERS arrays through galvanic replacement of silver onto electrochemically deposited copper micropatterns.
Facile fabrication of SERS arrays through galvanic replacement of silver onto electrochemically deposited copper micropatterns.
Chemphyschem : a European journal of chemical physics and physical chemistry (2012-09-28)
Xi Ke, Baoping Lu, Jinhui Hao, Jing Zhang, Haiyan Qiao, Zhe Zhang, Chunyan Xing, Wenshu Yang, Bailin Zhang, Jilin Tang
ZUSAMMENFASSUNG
Scatter me: A fast and cost-effective approach for the fabrication of surface-enhanced Raman scattering (SERS) arrays is developed. The method applied combines microcontact printing, electrodeposition, and galvanic replacement without the need for expensive instruments and intricate processing. The as-prepared arrays show excellent SERS activity and high reproducibility for Rhodamine 6G.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Rhodamin 6G, Dye content ~95 %
Sigma-Aldrich
Rhodamin 6G, Dye content 99 %
Sigma-Aldrich
Rhodamin 6G, suitable for fluorescence, BioReagent
Sigma-Aldrich
Rhodamin 6G -perchlorat, Dye content 99 %
Supelco
Rhodamin 6G, analytical standard