Merck
  • Determination of limonin in dog plasma by liquid chromatography-tandem mass spectrometry and its application to a pharmacokinetic study.
Determination of limonin in dog plasma by liquid chromatography-tandem mass spectrometry and its application to a pharmacokinetic study.
Biomedical chromatography : BMC (2012-09-29)
Shi-jia Liu, Ling Zhou, Jun Zhang, Bo-yang Yu, Chang-yin Li, Zi-xiu Liu, Wen-zheng Ju
ZUSAMMENFASSUNG
A highly sensitive liquid chromatography-tandem mass spectrometry method was developed and validated for the determination of limonin in beagle dog plasma using nimodipine as internal standard. The analyte and internal standard (IS) were extracted with ether followed by a rapid isocratic elution with 10 mm ammonium acetate buffer-methanol (26:74, v/v) on a C18 column (150 × 2.1 mm i.d.) and subsequent analysis by mass spectrometry in the multiple reaction monitoring mode. The precursor to product ion transitions of m/z 469.4 → 229.3 and m/z 417.2 → 122.0 were used to measure the analyte and the IS. The assay was linear over the concentration range of 0.625-100 ng/mL for limonin in dog plasma. The lower limit of quantification was 0.312 ng/mL and the extraction recovery was >90.4% for limonin. The inter- and intra-day precision of the method at three concentrations was less than 9.9%. The method was successfully applied to pharmacokinetic study of limonin in dogs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ammoniumacetat, ACS reagent, ≥97%
Sigma-Aldrich
Ammoniumacetat, ≥99.99% trace metals basis
Supelco
Ammoniumacetat, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammoniumacetat, Molecular Biology, ≥98%
Sigma-Aldrich
Ammoniumacetat -Lösung, Molecular Biology, 7.5 M
Sigma-Aldrich
Ammoniumacetat, 99.999% trace metals basis
Sigma-Aldrich
Ammoniumacetat, reagent grade, ≥98%
Sigma-Aldrich
Ammoniumacetat, BioXtra, ≥98%
Sigma-Aldrich
Ammoniumacetat -Lösung, BioUltra, ~5 M in H2O
Sigma-Aldrich
Ammoniumacetat, BioUltra, ≥99.0%
Sigma-Aldrich
Limonin, from citrus seeds, >90% (HPLC)
Sigma-Aldrich
Nimodipin