Merck
Fluorescence photoswitching based on a photochromic pKa change in an aqueous solution.
Chemical communications (Cambridge, England) (2012-10-02)
Tuyoshi Fukaminato, Emi Tateyama, Nobuyuki Tamaoki
ZUSAMMENFASSUNG
Reversible fluorescence photoswitching of RSA-AZO dyad 1 was clearly demonstrated in an acidic aqueous solution. The fluorescence photoswitching mechanism is based on the reversible ring opening/closing reactions of the RSA unit induced by a photochromic pK(a) change along with the photoisomerization of the AZO unit.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Azobenzol, 98%