Merck
  • Interactions of sulforaphane and dimethyl sulfoxide with methyl methanesulfonate, urethane, 4-nitroquinoline-1-oxide and hydrogen peroxide in the Drosophila melanogaster wing spot test.
Interactions of sulforaphane and dimethyl sulfoxide with methyl methanesulfonate, urethane, 4-nitroquinoline-1-oxide and hydrogen peroxide in the Drosophila melanogaster wing spot test.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-10-03)
I E Dueñas-García, L F Santos-Cruz, L Castañeda-Partida, A N Castañeda-Sortibrán, M G Ordaz-Téllez, A Sánchez-Santos, A Durán-Díaz, R Rodríguez-Arnaiz, M E Heres-Pulido
ZUSAMMENFASSUNG
Sulforaphane (SF) is an isothiocyanate present in Brassicaceae, vegetables that induce the detoxification of electrophiles and reactive oxygen species. SF has been correlated with chemoprevention mechanisms against degenerative diseases. We tested if the SF had an effect against methyl methanesulfonate (MMS), urethane (URE), 4-NQO and H(2)O(2). SF (>95% purity, 0.14, 0.28, 0.56 mM) was diluted in a DMSO/Tw80/EtOH mixture (DTE) corresponding to 25, 50, 100% of lyophilized broccoli. The SF treatment (0.14 mM) was positive for small spots in the ST cross and negative in the HB cross. In the HB cross, SF (0.28 mM) was genotoxic. In the ST cross, the SF treatments showed a tendency to reduce the genotoxic damage caused by MMS, which could be explained by the radical scavenging action of the DTE mixture. In the ST cross, the frequency of small spots in the SF 0.14 mM/URE treatment was similar to that of Water/URE, which can be explained by a DTE and SF scavenger action. In both crosses, the results for the direct oxidants, 4-NQO and H(2)O(2), were different and must be related to differential modulation of CYPs expression and the SF and DTE scavenger properties.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Urethan, ≥99%
Sigma-Aldrich
4-Nitrochinolin N-Oxid, ≥98%
Sigma-Aldrich
Urethan, ≥99.0% (GC)
Supelco
Urethan, analytical standard