Merck
  • In vivo phosphoantigen levels in bisphosphonate-treated human breast tumors trigger Vγ9Vδ2 T-cell antitumor cytotoxicity through ICAM-1 engagement.
In vivo phosphoantigen levels in bisphosphonate-treated human breast tumors trigger Vγ9Vδ2 T-cell antitumor cytotoxicity through ICAM-1 engagement.
Clinical cancer research : an official journal of the American Association for Cancer Research (2012-10-04)
Ismahène Benzaïd, Hannu Mönkkönen, Edith Bonnelye, Jukka Mönkkönen, Philippe Clézardin
ZUSAMMENFASSUNG
Nitrogen-containing bisphosphonates (N-BP) such as zoledronate and risedronate exhibit antitumor effects. They block the activity of farnesyl pyrophosphate synthase (FPPS) in the mevalonate pathway, leading to intracellular accumulation of mevalonate metabolites (IPP/ApppI), which are recognized as tumor phosphoantigens by Vγ9Vδ2 T cells. However, mechanisms responsible for Vγ9Vδ2 T-cell recognition of N-BP-treated tumors producing IPP/ApppI remain unclear. The effects of N-BPs on Vγ9Vδ2 T-cell expansion and anticancer activity were evaluated in vitro and in animal models of human breast cancers. The modalities of recognition of breast tumors by Vγ9Vδ2 T cells in N-BP-treated animals were also examined. We found a strong correlation between Vγ9Vδ2 T-cell anticancer activity and intracellular accumulation of IPP/ApppI in risedronate-treated breast cancer cells in vitro. In addition, following risedronate treatment of immunodeficient mice bearing human breast tumors, human Vγ9Vδ2 T cells infiltrated and inhibited growth of tumors that produced high IPP/ApppI levels but not those expressing low IPP/ApppI levels. The combination of doxorubicin with a N-BP improved, however, Vγ9Vδ2 T-cell cytotoxicity against breast tumors expressing low IPP/ApppI levels. Moreover, Vγ9Vδ2 T-cell cytotoxicity in mice treated with risedronate or zoledronate did not only depend on IPP/ApppI accumulation in tumors but also on expression of tumor cell surface receptor intercellular adhesion molecule-1 (ICAM-1), which triggered the recognition of N-BP-treated breast cancer cells by Vγ9Vδ2 T cells in vivo. These findings suggest that N-BPs can have an adjuvant role in cancer therapy by activating Vγ9Vδ2 T-cell cytotoxicity in patients with breast cancer that produces high IPP/ApppI levels after N-BP treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Etidronsäure, 60% aqueous solution
Sigma-Aldrich
Etidronate disodium hydrate, ≥97% (NMR), solid