Merck
  • [Role of allelopathic compositions in the regulation and development of legume-rhizobial symbiosis].
[Role of allelopathic compositions in the regulation and development of legume-rhizobial symbiosis].
Prikladnaia biokhimiia i mikrobiologiia (2012-10-06)
L E Makarova, V I Smirnov, L V Klyba, I G Petrova, L V Dudareva
ZUSAMMENFASSUNG
It was discovered that aromatic compounds isolated from root exudates of three legume species (Pisum sativum L., Vicia faba L. var. major Hartz, and Glycine max L. MERR) and identified as N-phenyl-2-naphthyl amine, dibutyl, and dioctyl esters of orthophthalic acid, which are known to work as negative allelopathic substances, are involved in the regulation of legume-rhizobial symbiosis formation after the inoculation of roots with rhizobia under unfavorable conditions for symbiosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumhydrogenphthalat, BioXtra, ≥99.95%
Supelco
Kaliumphthalat, einbasisch, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Kaliumphthalat, einbasisch, ACS reagent, acidimetric standard
Sigma-Aldrich
Phthalsäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Kaliumphthalat, einbasisch, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
N-Phenyl-2-naphthylamin, 95%
Supelco
2-Naphthylamin, analytical standard
Sigma-Aldrich
Kaliumphthalat, einbasisch, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Phthalsäure, puriss. p.a., ≥99.5% (T)
Supelco
Kaliumphthalat, einbasisch, qNMR standard, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Phthalsäure, reagent grade, 98%