Merck
  • 11β-Hydroxysteroid dehydrogenase type 2 in zebrafish brain: a functional role in hypothalamus-pituitary-interrenal axis regulation.
11β-Hydroxysteroid dehydrogenase type 2 in zebrafish brain: a functional role in hypothalamus-pituitary-interrenal axis regulation.
The Journal of endocrinology (2012-10-09)
Sarah L Alderman, Mathilakath M Vijayan
ZUSAMMENFASSUNG
The type 2, 11β-hydroxysteroid dehydrogenase (Hsd11b2) converts active glucocorticoids to their inactive derivatives (e.g. cortisol to cortisone). In most vertebrates, Hsd11b2 is essential for conferring aldosterone-specific actions in mineralocorticoid target tissues and for protecting glucocorticoid-sensitive tissues during stress. However, teleosts do not synthesize aldosterone, and the function of Hsd11b2 is poorly defined. The distribution of Hsd11b2 in nonmammalian brain is also largely unexplored. We tested the hypothesis that modulation of brain Hsd11b2 activity is involved in stressor-mediated cortisol regulation in zebrafish (Danio rerio). In adult zebrafish, the stress effect on Hsd11b2 expression in the brain was tested using acute air exposure followed by recovery over a 24-h period. hsd11b2 transcripts were found in nearly all peripheral tissues examined, and a spatial map of its mRNA abundance in unstressed zebrafish brain revealed extensive distribution. Stressor exposure increased the conversion of (3)H-cortisol to (3)H-cortisone indicating enhanced Hsd11b2 activity in zebrafish brain. Promoter analysis of zebrafish hsd11b2 gene revealed putative sites for cortisol-mediated transcriptional regulation of this gene. Furthermore, inhibition of Hsd11b2 activity by 18β-glycyrrhetinic acid resulted in elevated whole-body cortisol levels and preoptic area mRNA abundance of corticotropin-releasing factor and mineralocorticoid receptor. Taken together, our results underscore an important role for brain Hsd11b2 involvement in the negative feedback regulation of cortisol poststress in zebrafish.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
18β-Glycyrrhetinsäure, 97%
Sigma-Aldrich
18α-Glycyrrhetinsäure, ≥95%