Merck
  • Cyclobotryoxide, a phytotoxic metabolite produced by the plurivorous pathogen Neofusicoccum australe.
Cyclobotryoxide, a phytotoxic metabolite produced by the plurivorous pathogen Neofusicoccum australe.
Journal of natural products (2012-10-11)
Anna Andolfi, Lucia Maddau, Alessio Cimmino, Benedetto T Linaldeddu, Antonio Franceschini, Salvatorica Serra, Sara Basso, Dominique Melck, Antonio Evidente
ZUSAMMENFASSUNG
Two isolates of Neofusicoccum australe belonging to ITS haplotypes H4 and H1 and associated with grapevine cordon dieback and branch dieback of Phoenicean juniper, respectively, have been shown to produce in vitro structurally different secondary metabolites. From the strain BOT48 of N. australe (haplotype H4) a new cyclohexenone oxide, namely, cyclobotryoxide, was isolated together with 3-methylcatechol and tyrosol. Cyclobotryoxide was characterized as (1S,5R,6S)-5-hydroxy-3-methoxy-4-methyl-7-oxabicyclo[4.1.0]hept-3-en-2-one by spectroscopic, optical, and chemical methods. The strain BL24 (haplotype H1) produced tyrosol along with botryosphaerone D and (3S,4S)-3,4,8-trihydroxy-6-methoxy-3,4-dihydro-1(2H)-naphthalenone. The metabolites obtained from both strains were tested at four concentrations on leaves of grapevine cv. Cannonau, holm oak, and cork oak by the leaf puncture assay. Cyclobotryoxide proved to be the most phytotoxic compound. Tyrosol and cyclobotryoxide were also tested on detached grapevine leaves at concentrations of 0.25 and 0.5 mg/mL. Only cyclobotryoxide was found to be active in this bioassay.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-Methylcatechin, 98%