Merck
  • Metabolic disposition of the insect repellent DEET and the sunscreen oxybenzone following intravenous and skin administration in rats.
Metabolic disposition of the insect repellent DEET and the sunscreen oxybenzone following intravenous and skin administration in rats.
International journal of toxicology (2012-10-16)
Daryl J Fediuk, Tao Wang, Yufei Chen, Fiona E Parkinson, Michael P Namaka, Keith J Simons, Frank J Burczynski, Xiaochen Gu
ZUSAMMENFASSUNG
Insect repellent N,N-diethyl-m-toluamide (DEET) and sunscreen oxybenzone have shown a synergistic percutaneous enhancement when applied concurrently. Both compounds are extensively metabolized in vivo into a series of potentially toxic metabolites: 2 metabolites of DEET, N,N-diethyl-m-hydroxymethylbenzamide (DHMB) and N-ethyl-m-toluamide (ET), and 3 metabolites of oxybenzone, 2,4-dihydroxybenzophenone (DHB), 2,2-dihydroxy-4-methoxybenzophenone (DMB), and 2,3,4-trihydroxybenzophenone (THB). In this study, the metabolites were extensively distributed following intravenous and topical skin administration of DEET and oxybenzone in rats. Combined application enhanced the disposition of all DEET metabolites in the liver but did not consistently affect the distribution of oxybenzone metabolites. The DHMB appeared to be the major metabolite for DEET, while THB and its precursor DHB were the main metabolites for oxybenzone. Repeated once-daily topical application for 30 days led to higher concentrations of DEET metabolites in the liver. Hepatoma cell studies revealed a decrease in cellular proliferation from all metabolites as single and combined treatments, most notably at 72 hours. Increased accumulation of DHMB and ET in the liver together with an ability to reduce cellular proliferation at achievable plasma concentrations indicated that simultaneous exposure to DEET and oxybenzone might have the potential to precipitate adverse effects in a rat animal model.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Hydroxy-4-Methoxybenzophenon, 98%
Sigma-Aldrich
N,N-Diethyl-3-Methylbenzamid, 97%
Supelco
DEET, PESTANAL®, analytical standard
Supelco
Oxybenzon, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Oxybenzon, analytical standard